missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-proline Methyl Ester 97.0+%, TCI America™

Catalog No. p-7124017
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52285G 5 g
B522825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52285G Supplier TCI America Supplier No. B52285G
Only null left

Chemical Identifiers

CAS 59936-29-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01075092
InChI Key WVDGSSCWFMSRHN-QMMMGPOBSA-N
Synonym boc-l-pro-ome, boc-pro-ome, boc-l-proline-methyl ester, s-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, boc-l-proline methyl ester, 1-tert-butyl 2-methyl 2s-pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl s-pyrrolidine-1,2-dicarboxylate, pubchem18390, boc-l-prolinemethylester, boc-l-prolinem ethylester
PubChem CID 1277666
IUPAC Name 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
SMILES COC(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 229.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-proline Methyl Ester
TSCA No
Recommended Storage Refrigerator