accessibility menu, dialog, popup

UserName

CH3OO(S)NOOH3CCH3H3C

CAS RN 59936-29-7

IUPAC Name: 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: WVDGSSCWFMSRHN-QMMMGPOBSA-N
SMILES: COC(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)