missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-methioninol 95.0+%, TCI America™

Catalog No. B42835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42835G 5 g
B42831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42835G Supplier TCI America Supplier No. B42835G

Chemical Identifiers

CAS 51372-93-1
Molecular Formula C10H21NO3S
Molecular Weight (g/mol) 235.342
MDL Number MFCD00235939
InChI Key IPIBDQMAIDPJBU-QMMMGPOBSA-N
Synonym N-Boc-L-methioninol, (S)-2-(tert-Butoxycarbonyl)amino-4-methylthio-1-butanol, (S)-2-(Boc-amino)-4-methylthio-1-butanol
PubChem CID 10988238
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-4-methylsulfanylbutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CCSC)CO

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.34
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-methioninol