CAS RN 51372-93-1
CAS RN 51372-93-1
IUPAC Name:
tert-butyl N-[1-hydroxy-4-(methylsulfanyl)butan-2-yl]carbamate
Synonyms:
tert-butyl N-[1-hydroxy-4-(methylsulfanyl)butan-2-yl]carbamate
Molecular Weight (g/mol):
235.34
Molecular Formula:
C10H21NO3S
InChi Key:
IPIBDQMAIDPJBU-UHFFFAOYNA-N
SMILES:
CSCCC(CO)NC(=O)OC(C)(C)C
1
–
1
of
1
results
1
N-(tert-Butoxycarbonyl)-L-methioninol 95.0+%, TCI America™
CAS: 51372-93-1 Molecular Formula: C10H21NO3S Molecular Weight (g/mol): 235.342 MDL Number: MFCD00235939 InChI Key: IPIBDQMAIDPJBU-QMMMGPOBSA-N Synonym: N-Boc-L-methioninol, (S)-2-(tert-Butoxycarbonyl)amino-4-methylthio-1-butanol, (S)-2-(Boc-amino)-4-methylthio-1-butanol PubChem CID: 10988238 IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-4-methylsulfanylbutan-2-yl]carbamate SMILES: CC(C)(C)OC(=O)NC(CCSC)CO