missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-leucine Monohydrate 99.0+%, TCI America™
$50.33 - $123.03
Chemical Identifiers
| CAS | 13139-15-6 |
|---|---|
| Molecular Formula | C11H21NO4 |
| Molecular Weight (g/mol) | 231.29 |
| MDL Number | MFCD00066067 |
| InChI Key | MDXGYYOJGPFFJL-SVGMAFHSNA-N |
| Synonym | boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine |
| PubChem CID | 83170 |
| ChEBI | CHEBI:28702 |
| IUPAC Name | (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid |
| SMILES | CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Chemical Identifiers
| 13139-15-6 | |
| 231.29 | |
| MDXGYYOJGPFFJL-SVGMAFHSNA-N | |
| 83170 | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid |
| C11H21NO4 | |
| MFCD00066067 | |
| boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
| CHEBI:28702 | |
| CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Specifications
| 13139-15-6 | |
| White | |
| MFCD00066067 | |
| boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
| CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 231.29 | |
| CHEBI:28702 | |
| ≥99.0% (T) | |
| N-(tert-Butoxycarbonyl)-L-leucine Monohydrate |
| 87°C | |
| C11H21NO4 | |
| 5 g | |
| MDXGYYOJGPFFJL-SVGMAFHSNA-N | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid | |
| 83170 | |
| 231.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes