missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-leucine Monohydrate 99.0+%, TCI America™
Supplier: TCI America B118725G
Specifications
| N-(tert-Butoxycarbonyl)-L-leucine Monohydrate | |
| 87°C | |
| C11H21NO4 | |
| 25 g | |
| MDXGYYOJGPFFJL-SVGMAFHSNA-N | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid | |
| 83170 | |
| 231.29 | |
| Crystalline Powder |
| 13139-15-6 | |
| White | |
| MFCD00066067 | |
| boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
| CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 231.29 | |
| CHEBI:28702 | |
| ≥99.0% (T) |
Chemical Identifiers
| 13139-15-6 | |
| 231.29 | |
| MDXGYYOJGPFFJL-SVGMAFHSNA-N | |
| 83170 | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid |
| C11H21NO4 | |
| MFCD00066067 | |
| boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
| CHEBI:28702 | |
| CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Safety and Handling
TSCA : Yes