missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-isoleucine Hemihydrate 99.0+%, TCI America™

Catalog No. p-7118904
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B11865G 5 g
B118625G 25 g
2 options

Catalog No. B11865G

Supplier: TCI America B11865G

Only null left

Chemical Identifiers

CAS 13139-16-7
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038324 MFCD00237550
InChI Key QJCNLJWUIOIMMF-YUMQZZPRSA-N
Synonym boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine
PubChem CID 2724762
IUPAC Name (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 231.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-isoleucine Hemihydrate