accessibility menu, dialog, popup

UserName

CH3(S)(S)HNOOCH3H3CCH3OOHH3C

CAS RN 13139-16-7

IUPAC Name: (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoic acid
Synonyms: (2S,3S)-2-[(tert-butoxycarbonyl)amino]-3-methylpentanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: QJCNLJWUIOIMMF-YUMQZZPRSA-N
SMILES: CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)