missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-homoserine 98.0+%, TCI America™

Catalog No. B40255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40255G 5 g
B40251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40255G Supplier TCI America Supplier No. B40255G

Chemical Identifiers

CAS 41088-86-2
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00057839
InChI Key PZEMWPDUXBZKJN-LURJTMIESA-N
Synonym N-Boc-L-homoserine, (S)-2-(tert-Butoxycarbonylamino)-4-hydroxybutyric Acid, (S)-2-(Boc-amino)-4-hydroxybutyric Acid
PubChem CID 13066457
IUPAC Name (2S)-4-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCO)C(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 219.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-homoserine
TSCA No
Recommended Storage Freezer