accessibility menu, dialog, popup

UserName

CAS RN 41088-86-2

CH3CH3H3COOHNHOOOH

CAS RN 41088-86-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-hydroxybutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-hydroxybutanoic acid
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: PZEMWPDUXBZKJN-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCO)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results