missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-2-cyclohexylglycine 98.0+%, TCI America™
$41.77 - $91.13
Chemical Identifiers
| CAS | 109183-71-3 |
|---|---|
| Molecular Formula | C13H23NO4 |
| Molecular Weight (g/mol) | 257.33 |
| MDL Number | MFCD00133638 |
| InChI Key | QSUXZIPXYDQFCX-JTQLQIEISA-N |
| Synonym | boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid |
| PubChem CID | 7004938 |
| IUPAC Name | (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
| SMILES | CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O |
Chemical Identifiers
| 109183-71-3 | |
| 257.33 | |
| QSUXZIPXYDQFCX-JTQLQIEISA-N | |
| 7004938 | |
| CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O |
| C13H23NO4 | |
| MFCD00133638 | |
| boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid | |
| (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
Specifications
| 83°C | |
| White | |
| MFCD00133638 | |
| QSUXZIPXYDQFCX-JTQLQIEISA-N | |
| (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid | |
| 7004938 | |
| ≥98.0% (HPLC) | |
| N-(tert-Butoxycarbonyl)-L-2-cyclohexylglycine |
| 109183-71-3 | |
| C13H23NO4 | |
| boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid | |
| CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O | |
| 257.33 | |
| 257.33 | |
| Crystalline Powder |
Safety and Handling
TSCA : No