missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-2-cyclohexylglycine 98.0+%, TCI America™

Catalog No. B38391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38391G 1 g
B38395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38391G Supplier TCI America Supplier No. B38391G

Chemical Identifiers

CAS 109183-71-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD00133638
InChI Key QSUXZIPXYDQFCX-JTQLQIEISA-N
Synonym boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
PubChem CID 7004938
IUPAC Name (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Melting Point 83°C
Color White
Quantity 1 g
Formula Weight 257.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-2-cyclohexylglycine