missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)glycine tert-Butyl Ester 98.0+%, TCI America™

Catalog No. B41561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41561G 1 g
B41565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41561G Supplier TCI America Supplier No. B41561G

Chemical Identifiers

CAS 111652-20-1
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00191866
InChI Key ZDKFMHKWZATBMR-UHFFFAOYSA-N
Synonym boc-gly-otbu, tert-butyl 2-tert-butoxycarbonyl amino acetate, boc-glycine tert-butyl ester, n-boc-glycine tert-butyl ester, n-tert-butoxycarbonyl glycine tert-butyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, 1,1-dimethylethyl ester, tert-butyl 2-2-methylpropan-2-yl oxycarbonylamino acetate, 2-azabutanedioic acid di-tert-butyl ester
PubChem CID 4097102
IUPAC Name tert-butyl 2-{[(tert-butoxy)carbonyl]amino}acetate
SMILES CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C

Specifications

Melting Point 67°C
Color White
Quantity 1 g
Formula Weight 231.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)glycine tert-Butyl Ester