accessibility menu, dialog, popup

UserName

CH3CH3H3COONHOOCH3CH3H3C

CAS RN 111652-20-1

IUPAC Name: tert-butyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms: tert-butyl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: ZDKFMHKWZATBMR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)