missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)glycine Ethyl Ester 98.0+%, TCI America™

Catalog No. B47825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47825G 5 g
B478225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47825G Supplier TCI America Supplier No. B47825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14719-37-0
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD05663720
InChI Key CNIBHMMDDXGDNR-UHFFFAOYSA-N
Synonym n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
PubChem CID 357729
IUPAC Name ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CCOC(=O)CNC(=O)OC(C)(C)C

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)glycine Ethyl Ester