missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 93.0+%, TCI America™

Catalog No. B32711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32711G 1 g
B32715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32711G Supplier TCI America Supplier No. B32711G
Only null left

Chemical Identifiers

CAS 67341-01-9
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD01883039
InChI Key IBDIOGYTZBKRGI-UHFFFAOYSA-N
Synonym n-boc-dl-phenylglycinol, tert-butyl 2-hydroxy-1-phenylethyl carbamate, 2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl n-2-hydroxy-1-phenylethyl carbamate, boc-l-phenylglycinol, carbamic acid, 2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol, n-boc-l-2-phenylglycinol, pubchem13817, acmc-209uhj
PubChem CID 5050210
IUPAC Name tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate
SMILES CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 237.30
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol