accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNOH

CAS RN 67341-01-9

IUPAC Name: tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate
Synonyms: tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: IBDIOGYTZBKRGI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results