missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-valine 98.0+%, TCI America™

Catalog No. B29915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29915G 5 g
B299125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29915G Supplier TCI America Supplier No. B29915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22838-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00038282
InChI Key SZXBQTSZISFIAO-SSDOTTSWSA-N
Synonym boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine
PubChem CID 637605
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-valine