accessibility menu, dialog, popup

UserName

H3CCH3(R)NHOOCH3CH3H3COHO

CAS RN 22838-58-0

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-methylbutanoic acid|BOC-D-VAL-OH
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: SZXBQTSZISFIAO-SSDOTTSWSA-N
SMILES: CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)