missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-tyrosine 98.0+%, TCI America™

Catalog No. B29631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29631G 1 g
B29635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29631G Supplier TCI America Supplier No. B29631G

Chemical Identifiers

CAS 70642-86-3
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.31
MDL Number MFCD00063030
InChI Key CNBUSIJNWNXLQQ-LDGXTIHJNA-N
Synonym boc-d-tyrosine, boc-d-tyr-oh, n-boc-d-tyrosine, n-tert-butoxycarbonyl-d-tyrosine, d-tyrosine, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butoxycarbonyl-d-tyrosine, n-alpha-t-butyloxycarbonyl-d-tyrosine, r-2-tert-butoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid, r-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoic acid, 2r-3-4-hydroxyphenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
PubChem CID 1549481
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 281.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-tyrosine