accessibility menu, dialog, popup

UserName

HN(S)OOOOHHOCH3CH3CH3

CAS RN 70642-86-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 281.31
Molecular Formula: C14H19NO5
InChi Key: CNBUSIJNWNXLQQ-LDGXTIHJNA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results