missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-serine 98.0+%, TCI America™

Catalog No. B22581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22581G 1 g
B22585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22581G Supplier TCI America Supplier No. B22581G

Chemical Identifiers

CAS 6368-20-3
Molecular Formula C8H15NO5
Molecular Weight (g/mol) 205.21
MDL Number MFCD00063142
InChI Key FHOAKXBXYSJBGX-RXMQYKEDSA-N
Synonym boc-d-serine, boc-d-ser-oh, n-boc-d-serine, n-tert-butoxycarbonyl-d-serine, n-alpha-t-butyloxycarbonyl-d-serine, boc-l-serine, boc-l-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, d-serine, n-1,1-dimethylethoxy carbonyl, r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid
PubChem CID 637603
IUPAC Name (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)O

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 205.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-serine