missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-serine Methyl Ester 98.0+%, TCI America™
$39.87 - $131.00
Chemical Identifiers
| CAS | 95715-85-8 |
|---|---|
| Molecular Formula | C9H17NO5 |
| Molecular Weight (g/mol) | 219.237 |
| MDL Number | MFCD00270516 |
| InChI Key | SANNKFASHWONFD-ZCFIWIBFSA-N |
| Synonym | boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate |
| PubChem CID | 377723 |
| IUPAC Name | methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate |
| SMILES | CC(C)(C)OC(=O)NC(CO)C(=O)OC |
Chemical Identifiers
| 95715-85-8 | |
| 219.237 | |
| SANNKFASHWONFD-ZCFIWIBFSA-N | |
| 377723 | |
| CC(C)(C)OC(=O)NC(CO)C(=O)OC |
| C9H17NO5 | |
| MFCD00270516 | |
| boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate | |
| methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate |
Specifications
| 95715-85-8 | |
| Yellow | |
| MFCD00270516 | |
| SANNKFASHWONFD-ZCFIWIBFSA-N | |
| methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate | |
| 377723 | |
| ≥98.0% (HPLC,N) | |
| N-(tert-Butoxycarbonyl)-D-serine Methyl Ester |
| 215°C | |
| C9H17NO5 | |
| boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate | |
| CC(C)(C)OC(=O)NC(CO)C(=O)OC | |
| 219.237 | |
| 219.24 | |
| Liquid |
Safety and Handling
TSCA : No