missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-proline Succinimidyl Ester 98.0+%, TCI America™

Catalog No. B34145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34145G 5 g
B34141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34145G Supplier TCI America Supplier No. B34145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102185-34-2
Molecular Formula C14H20N2O6
Molecular Weight (g/mol) 312.32
MDL Number MFCD00069687
InChI Key DICWIJISMKZDDY-UHFFFAOYNA-N
Synonym boc-d-pro-osu, n-tert-butoxycarbonyl-d-proline succinimidyl ester, boc-d-proline n-hydroxysuccinimide ester, 1-tert-butyl 2-2,5-dioxopyrrolidin-1-yl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline succinimidyl ester, n-t-boc-d-proline n-hydroxysuccinimide*ester, n-tert-butoxycarbonyl-d-prolinesuccinimidylester, n-alpha-t-butyloxycarbonyl-d-proline succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 1-tert-butoxycarbonyl-d-prolinate, 1-o-tert-butyl 2-o-2,5-dioxopyrrolidin-1-yl 2r-pyrrolidine-1,2-dicarboxylate
PubChem CID 853884
IUPAC Name 1-tert-butyl 2-(2,5-dioxopyrrolidin-1-yl) pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)ON1C(=O)CCC1=O

Specifications

Color White
Quantity 5 g
Formula Weight 312.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-proline Succinimidyl Ester
TSCA No
Recommended Storage Freezer