accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOONOO

CAS RN 102185-34-2

IUPAC Name: 1-tert-butyl 2-(2,5-dioxopyrrolidin-1-yl) pyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-(2,5-dioxopyrrolidin-1-yl) pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 312.32
Molecular Formula: C14H20N2O6
InChi Key: DICWIJISMKZDDY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results