missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-methionine 95.0+%, TCI America™

Catalog No. p-7119867
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29675G 5 g
B296725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29675G Supplier TCI America Supplier No. B29675G
Only null left

Chemical Identifiers

CAS 5241-66-7
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
MDL Number MFCD00038256
InChI Key IMUSLIHRIYOHEV-SSDOTTSWSA-N
Synonym boc-d-met-oh, boc-d-methionine, n-boc-d-methionine, n-tert-butoxycarbonyl-d-methionine, n-tert-butyloxycarbonyl-d-methionine, boc-l-met-oh, d-methionine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-methionine, n-alpha-t-butyloxycarbonyl-d-methionine
PubChem CID 111051
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 249.33
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-methionine