accessibility menu, dialog, popup

UserName

H3CS(R)NHOOCH3CH3H3CHOO

CAS RN 5241-66-7

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-4-(methylsulfanyl)butanoic acid| BOC-D-MET-OH
Molecular Weight (g/mol): 249.33
Molecular Formula: C10H19NO4S
InChi Key: IMUSLIHRIYOHEV-SSDOTTSWSA-N
SMILES: CSCC[C@@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results