missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-2-phenylglycinol 98.0+%, TCI America™
$53.16 - $210.75
Chemical Identifiers
| CAS | 102089-74-7 |
|---|---|
| Molecular Formula | C13H19NO3 |
| Molecular Weight (g/mol) | 237.299 |
| MDL Number | MFCD00274205 |
| InChI Key | IBDIOGYTZBKRGI-NSHDSACASA-N |
| Synonym | boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol |
| PubChem CID | 7016461 |
| IUPAC Name | tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 |
Chemical Identifiers
| 102089-74-7 | |
| 237.299 | |
| IBDIOGYTZBKRGI-NSHDSACASA-N | |
| 7016461 | |
| CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 |
| C13H19NO3 | |
| MFCD00274205 | |
| boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol | |
| tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate |
Specifications
| 137°C | |
| Yellow | |
| MFCD00274205 | |
| IBDIOGYTZBKRGI-NSHDSACASA-N | |
| tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate | |
| 7016461 | |
| ≥98.0% (GC) | |
| N-(tert-Butoxycarbonyl)-D-2-phenylglycinol |
| 102089-74-7 | |
| C13H19NO3 | |
| boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol | |
| CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 | |
| 237.299 | |
| 237.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No