missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-2-phenylglycinol 98.0+%, TCI America™

Catalog No. B32705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32705G 5 g
B32701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32705G Supplier TCI America Supplier No. B32705G

Chemical Identifiers

CAS 102089-74-7
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD00274205
InChI Key IBDIOGYTZBKRGI-NSHDSACASA-N
Synonym boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol
PubChem CID 7016461
IUPAC Name tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1

Specifications

Melting Point 137°C
Color Yellow
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-2-phenylglycinol