missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-beta-alanine 98.0+%, TCI America™
$43.67 - $66.46
Chemical Identifiers
| CAS | 3303-84-2 |
|---|---|
| Molecular Formula | C8H15NO4 |
| Molecular Weight (g/mol) | 189.21 |
| MDL Number | MFCD00037291 |
| InChI Key | WCFJUSRQHZPVKY-UHFFFAOYSA-N |
| Synonym | boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid |
| PubChem CID | 76809 |
| IUPAC Name | 3-{[(tert-butoxy)carbonyl]amino}propanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCC(O)=O |
Chemical Identifiers
| 3303-84-2 | |
| 189.21 | |
| WCFJUSRQHZPVKY-UHFFFAOYSA-N | |
| 76809 | |
| CC(C)(C)OC(=O)NCCC(O)=O |
| C8H15NO4 | |
| MFCD00037291 | |
| boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid | |
| 3-{[(tert-butoxy)carbonyl]amino}propanoic acid |
Specifications
| 77°C | |
| White | |
| MFCD00037291 | |
| WCFJUSRQHZPVKY-UHFFFAOYSA-N | |
| 3-{[(tert-butoxy)carbonyl]amino}propanoic acid | |
| 189.21 | |
| 189.21 | |
| Crystalline Powder |
| 3303-84-2 | |
| C8H15NO4 | |
| boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid | |
| CC(C)(C)OC(=O)NCCC(O)=O | |
| 5 g | |
| 76809 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-beta-alanine |
Safety and Handling
TSCA : No