missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-oxo-L-proline 98.0+%, TCI America™

Catalog No. p-7123374
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B41411G 1 g
B41415G 5 g
2 options

Catalog No. B41411G

Supplier: TCI America B41411G

Only null left

Chemical Identifiers

CAS 84348-37-8
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD01860669
InChI Key CKYGSXRXTIKGAJ-UHFFFAOYNA-N
Synonym n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
PubChem CID 11593804
IUPAC Name 1-[(tert-butoxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 229.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-oxo-L-proline
TSCA No
Recommended Storage Refrigerator