accessibility menu, dialog, popup

UserName

CAS RN 84348-37-8

CH3H3CCH3OONOOHO

CAS RN 84348-37-8

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 229.23
Molecular Formula: C10H15NO5
InChi Key: CKYGSXRXTIKGAJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results