missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-nitro-L-phenylalanine 98.0+%, TCI America™

Catalog No. p-7119568
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22321G 1 g
B22325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22321G Supplier TCI America Supplier No. B22321G
Only null left

Chemical Identifiers

CAS 33305-77-0
Molecular Formula C14H18N2O6
Molecular Weight (g/mol) 310.306
MDL Number MFCD00038128
InChI Key XBQADBXCNQPHHY-NSHDSACASA-N
Synonym boc-4-nitro-l-phenylalanine, boc-phe 4-no2-oh, boc-l-4-nitrophenylalanine, boc-l-4-nitrophe, n-boc-4-nitro-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, boc-l-4-no2-phe-oh, 2s-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-l-phenylalanine, 2s-2-tert-butoxy carbonyl amino-3-4-nitrophenyl propanoic acid
PubChem CID 7021882
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 310.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-nitro-L-phenylalanine