accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNOOOHO

CAS RN 33305-77-0

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(4-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 310.31
Molecular Formula: C14H18N2O6
InChi Key: XBQADBXCNQPHHY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)