missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine 98.0+%, TCI America™

Catalog No. B36145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36145G 5 g
B36141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36145G Supplier TCI America Supplier No. B36145G

Chemical Identifiers

CAS 68090-88-0
Molecular Formula C14H17ClNO4
Molecular Weight (g/mol) 298.74
MDL Number MFCD00057808
InChI Key BETBOAZCLSJOBQ-NSHDSACASA-M
Synonym boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
PubChem CID 2734499
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoate
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C=C1)C([O-])=O

Specifications

Melting Point 112°C
Color White
Quantity 5 g
Formula Weight 299.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine