accessibility menu, dialog, popup

UserName

CAS RN 68090-88-0

H3CH3CCH3OONH(S)ClOO

CAS RN 68090-88-0

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-chlorophenyl)propanoate
Molecular Weight (g/mol): 298.74
Molecular Formula: C14H17ClNO4
InChi Key: BETBOAZCLSJOBQ-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results