missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-3-iodo-L-alanine Methyl Ester 98.0+%, TCI America™

Catalog No. p-7123314
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40441G 1 g
B40445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40441G Supplier TCI America Supplier No. B40441G
Only null left

Chemical Identifiers

CAS 93267-04-0
Molecular Formula C9H16INO4
Molecular Weight (g/mol) 329.13
MDL Number MFCD00216579
InChI Key UGZBFCCHLUWCQI-LURJTMIESA-N
Synonym boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
PubChem CID 10903591
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
SMILES COC(=O)[C@H](CI)NC(=O)OC(C)(C)C

Specifications

Melting Point 51°C
Color Yellow
Quantity 1 g
Formula Weight 329.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-iodo-L-alanine Methyl Ester
TSCA No
Recommended Storage Refrigerator