accessibility menu, dialog, popup

UserName

CAS RN 93267-04-0

IUPAC Name: methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
Synonyms: methyl (2R)-2-[(tert-butoxycarbonyl)amino]-3-iodopropanoate
Molecular Weight (g/mol): 329.13
Molecular Formula: C9H16INO4
InChi Key: UGZBFCCHLUWCQI-LURJTMIESA-N
SMILES: COC(=O)[C@H](CI)NC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)