missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 98.0+%, TCI America™
$69.31 - $207.92
Chemical Identifiers
| CAS | 114873-01-7 |
|---|---|
| Molecular Formula | C14H18FNO4 |
| Molecular Weight (g/mol) | 283.299 |
| MDL Number | MFCD00672522 |
| InChI Key | FPCCREICRYPTTL-NSHDSACASA-N |
| Synonym | boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine |
| PubChem CID | 2734492 |
| IUPAC Name | (2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O |
Chemical Identifiers
| 114873-01-7 | |
| 283.299 | |
| FPCCREICRYPTTL-NSHDSACASA-N | |
| 2734492 | |
| CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O |
| C14H18FNO4 | |
| MFCD00672522 | |
| boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine | |
| (2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 114873-01-7 | |
| White | |
| MFCD00672522 | |
| boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine | |
| CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O | |
| 283.299 | |
| 283.30 | |
| Crystalline Powder |
| 82°C | |
| C14H18FNO4 | |
| 1 g | |
| FPCCREICRYPTTL-NSHDSACASA-N | |
| (2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 2734492 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine |
Safety and Handling
TSCA : No