missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 98.0+%, TCI America™

Catalog No. B43671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43671G 1 g
B43675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43671G Supplier TCI America Supplier No. B43671G

Chemical Identifiers

CAS 114873-01-7
Molecular Formula C14H18FNO4
Molecular Weight (g/mol) 283.299
MDL Number MFCD00672522
InChI Key FPCCREICRYPTTL-NSHDSACASA-N
Synonym boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine
PubChem CID 2734492
IUPAC Name (2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 283.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine