missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide 98.0+%, TCI America™

Catalog No. p-7119609
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23031G 1 g
B23035G 5 g
B230325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B23031G Supplier TCI America Supplier No. B23031G
Only null left

Chemical Identifiers

CAS 198572-71-3
Molecular Formula C11H14N2O6S
Molecular Weight (g/mol) 302.30
MDL Number MFCD06796217
InChI Key WAYBGHIVTXTUCZ-UHFFFAOYSA-N
Synonym N-Boc-2-nitrobenzenesulfonamide
PubChem CID 11141172
IUPAC Name tert-butyl N-(2-nitrobenzenesulfonyl)carbamate
SMILES CC(C)(C)OC(=O)NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 302.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide