missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-2,2-dimethyl-1,3-propanediamine 98.0+%, TCI America™

Catalog No. B30521G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30521G 1 g
B30525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30521G Supplier TCI America Supplier No. B30521G
Only null left

Chemical Identifiers

CAS 292606-35-0
Molecular Formula C10H22N2O2
Molecular Weight (g/mol) 202.298
MDL Number MFCD01076212
InChI Key QLPCQDZWRXWROH-UHFFFAOYSA-N
Synonym N-Boc-2,2-dimethyl-1,3-propanediamine, tert-Butyl (3-Amino-2,2-dimethylpropyl)carbamate, (3-Amino-2,2-dimethylpropyl)carbamic Acid tert-Butyl Ester, N-Boc-2,2-dimethyl-1,3-diaminopropane
PubChem CID 4114359
IUPAC Name tert-butyl N-(3-amino-2,2-dimethylpropyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(C)(C)CN

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Formula Weight 202.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-2,2-dimethyl-1,3-propanediamine