accessibility menu, dialog, popup

UserName

CAS RN 292606-35-0

CH3CH3H3COONHCH3H3CH2N

CAS RN 292606-35-0

IUPAC Name: tert-butyl N-(3-amino-2,2-dimethylpropyl)carbamate
Synonyms: tert-butyl N-(3-amino-2,2-dimethylpropyl)carbamate
Molecular Weight (g/mol): 202.3
Molecular Formula: C10H22N2O2
InChi Key: QLPCQDZWRXWROH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(C)(C)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)