missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butoxycarbonyl-1,6-hexanediamine, 95%

Catalog No. AC301830010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301830010 1 g Plastic Bottle
AC301830050 5 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC301830010 Supplier Thermo Scientific Chemicals Supplier No. 301830010

CAS: 51857-17-1 | C11H24N2O2 | 216.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.33
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN

Specifications

Boiling Point 106°C to 110°C (0.3mmHg)
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic Bottle
Refractive Index 1.4590 to 1.461
Quantity 1 g
UN Number 2735
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,ch2cl2
Formula Weight 216.33
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material N-tert-Butoxycarbonyl-1, 6-hexanediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only