missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 95.0+%, TCI America™

Catalog No. B40815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40815G 5 g
B408125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40815G Supplier TCI America Supplier No. B40815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71026-66-9
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD00043022
InChI Key WIVYTYZCVWHWSH-UHFFFAOYSA-N
Synonym tert-butyl 4-aminophenyl carbamate, 4-tert-butoxycarbonylamino aniline, tert-butyl 4-aminophenylcarbamate, tert-butyl n-4-aminophenyl carbamate, n-boc-p-phenylenediamine, n-boc-1,4-phenylene diamine, n-boc-1,4-phenylenediamine, 4-amino-phenyl-carbamic acid tert-butyl ester, tert-butyl4-aminophenylcarbamate, 4-t-butyloxycarbonylamino aniline
PubChem CID 688611
IUPAC Name tert-butyl N-(4-aminophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)N

Specifications

Melting Point 116°C
Quantity 5 g
Formula Weight 208.26
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,4-phenylenediamine