missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,3-phenylenediamine 98.0+%, TCI America™

Catalog No. B42225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42225G 5 g
B42221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42225G Supplier TCI America Supplier No. B42225G

Chemical Identifiers

CAS 68621-88-5
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD06657915
InChI Key IEUIEMIRUXSXCL-UHFFFAOYSA-N
Synonym N-Boc-1,3-phenylenediamine, tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic Acid tert-Butyl Ester, 3-(tert-Butoxycarbonylamino)aniline, 3-(Boc-amino)aniline
PubChem CID 12877220
IUPAC Name tert-butyl N-(3-aminophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC(=C1)N

Specifications

Quantity 5 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,3-phenylenediamine