missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7123422
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42381G 1 g
B42385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42381G Supplier TCI America Supplier No. B42381G
Only null left

Chemical Identifiers

CAS 146651-75-4
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD02169707
InChI Key KCZFBLNQOSFGSH-UHFFFAOYSA-N
Synonym N-Boc-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, (2-Aminophenyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonylamino)aniline, 2-(Boc-amino)aniline
PubChem CID 676311
IUPAC Name tert-butyl N-(2-aminophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC=C1N

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,2-phenylenediamine