missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl N-methylcarbamate, 97%

CAS: 18342-66-0 | C6H8N2O4 | 172.14 g/mol

Supplier:  Thermo Scientific Chemicals H6356606

 View more versions of this product

Catalog No. AAH6356606


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

N-Succinimidyl N-methylcarbamate
C6H8N2O4
5 g
Moisture sensitive
Slightly soluble in water.
CNC(=O)ON1C(=O)CCC1=O
172.14
CHEBI:83949
97%
18342-66-0
MFCD00800292
1531863
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
XMNGSPOWUCNRMO-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
4381935
172.14
Solid

Chemical Identifiers

18342-66-0
172.14
XMNGSPOWUCNRMO-UHFFFAOYSA-N
4381935
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
C6H8N2O4
MFCD00800292
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
CHEBI:83949
CNC(=O)ON1C(=O)CCC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only