missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl N-methylcarbamate, 97%

Catalog No. p-7055077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6356603 1 g
AAH6356606 5 g
AAH6356614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6356603 Supplier Thermo Scientific Chemicals Supplier No. H6356603
Only null left

CAS: 18342-66-0 | C6H8N2O4 | 172.14 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 18342-66-0
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
MDL Number MFCD00800292
InChI Key XMNGSPOWUCNRMO-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
PubChem CID 4381935
ChEBI CHEBI:83949
IUPAC Name (2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
SMILES CNC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 1 g
Beilstein 1531863
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 172.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Succinimidyl N-methylcarbamate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only