missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl Methacrylate 98.0+%, TCI America™
Supplier: TCI America S081225G
Specifications
| N-Succinimidyl Methacrylate | |
| 104°C | |
| C8H9NO4 | |
| 25 g | |
| ACGJEMXWUYWELU-UHFFFAOYSA-N | |
| 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate | |
| 10130244 | |
| ≥98.0% (GC) |
| 38862-25-8 | |
| White | |
| MFCD15072193 | |
| Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide | |
| CC(=C)C(=O)ON1C(=O)CCC1=O | |
| 183.16 | |
| 183.16 | |
| Crystalline Powder |
Chemical Identifiers
| 38862-25-8 | |
| 183.16 | |
| ACGJEMXWUYWELU-UHFFFAOYSA-N | |
| 10130244 | |
| CC(=C)C(=O)ON1C(=O)CCC1=O |
| C8H9NO4 | |
| MFCD15072193 | |
| Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide | |
| 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator